Assay Details
| Gene Symbol: Slc35f5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028787.4 | NP_083063.2 | 11 - 12 | 1319 | 123 | |
| GenBank mRNA | AK004892.2 | - | 11 - 12 | 1300 | 123 | |
| AK076339.1 | - | 11 - 12 | 1313 | 123 | ||
| AK145848.1 | - | 11 - 12 | 1320 | 123 | ||
| AK169012.1 | - | 11 - 12 | 1181 | 123 | ||
| AK171951.1 | - | 11 - 12 | 1293 | 123 | ||
| BC026858.1 | - | 11 - 12 | 1178 | 4014064 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc35f5
Genomic Map