Assay Details
| Gene Symbol: Slc35f5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028787.4 | NP_083063.2 | 5 - 6 | 712 | 74 | |
| GenBank mRNA | AK004892.2 | - | 5 - 6 | 693 | 74 | |
| AK076339.1 | - | 5 - 6 | 706 | 74 | ||
| AK145848.1 | - | 5 - 6 | 713 | 74 | ||
| AK169012.1 | - | 5 - 6 | 574 | 74 | ||
| AK171951.1 | - | 5 - 6 | 686 | 74 | ||
| BC026858.1 | - | 5 - 6 | 571 | 4014064 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc35f5
Genomic Map