Assay Details
| Gene Symbol: Tom1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028011.2 | NP_082287.1 | 4 - 5 | 426 | 83 | |
| GenBank mRNA | AF395837.1 | - | 4 - 5 | 390 | 83 | |
| AJ505007.1 | - | 4 - 5 | 387 | 83 | ||
| AK033712.1 | - | 4 - 5 | 428 | 83 | ||
| AK083630.1 | - | 388 | 83 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tom1l1
Genomic Map