Assay Details
| Gene Symbol: Itfg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206335.1 | NP_001193264.1 | 6 - 7 | 1100 | 77 | |
| NM_207217.4 | NP_997100.1 | 5 - 6 | 988 | 77 | ||
| GenBank mRNA | AK029427.1 | - | 5 - 6 | 888 | 77 | |
| AK170646.1 | - | 6 - 7 | 1019 | 77 | ||
| BC046457.1 | - | 6 - 7 | 947 | 4222961 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itfg3
Genomic Map