Assay Details
| Gene Symbol: Itfg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206335.1 | NP_001193264.1 | 7 - 8 | 1234 | 110 | |
| NM_207217.4 | NP_997100.1 | 6 - 7 | 1122 | 110 | ||
| GenBank mRNA | AK029427.1 | - | 6 - 7 | 1022 | 110 | |
| AK158049.1 | - | 1216 | 110 | |||
| AK170646.1 | - | 7 - 8 | 1153 | 110 | ||
| BC046457.1 | - | 7 - 8 | 1081 | 4222961 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itfg3
Genomic Map