Assay Details
| Gene Symbol: Itfg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206335.1 | NP_001193264.1 | 11 - 12 | 1730 | 107 | |
| NM_207217.4 | NP_997100.1 | 10 - 11 | 1618 | 107 | ||
| GenBank mRNA | AK029427.1 | - | 10 - 11 | 1518 | 107 | |
| AK158049.1 | - | 1712 | 107 | |||
| AK170646.1 | - | 11 - 12 | 1649 | 107 | ||
| BC046457.1 | - | 11 - 12 | 1577 | 4222961 | 107 | |
More Information
Set Membership: |
|

Itfg3
Genomic Map