Assay Details
| Gene Symbol: Bnip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172149.5 | NP_742161.4 | 2 - 3 | 188 | 108 | |
| GenBank mRNA | AY547731.1 | - | 2 - 3 | 174 | 108 | |
| BC086888.1 | - | 2 - 3 | 177 | 30122069 | 108 | |
| CV560804.1 | - | 2 - 3 | 189 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bnip1
Genomic Map