Assay Details
| Gene Symbol: Capn15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079422.1 | - | 1 - 2 | 214 | 83 | |
| AK170543.1 | - | 1 - 2 | 214 | 83 | ||
| BC058094.1 | - | 1 - 2 | 239 | 6824892 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC058094.1 transcripts.
Set Membership: |
|

Capn15
Genomic Map