Assay Details
| Gene Symbol: Capn15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015830.1 | NP_056645.1 | 5 - 6 | 2224 | 84 | |
| GenBank mRNA | AK079422.1 | - | 4 - 5 | 2316 | 84 | |
| AK143711.1 | - | 7 - 8 | 2863 | 84 | ||
| AK148209.1 | - | 3 - 4 | 483 | 84 | ||
| AK170543.1 | - | 6 - 7 | 2458 | 84 | ||
| BC058094.1 | - | 6 - 7 | 2482 | 6824892 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Capn15
Genomic Map