Assay Details
Gene Symbol: Ergic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026170.3 | NP_080446.1 | 2 - 3 | 178 | 60 | |
GenBank mRNA | AB093571.1 | - | 2 - 3 | 77 | 60 | |
AK003239.1 | - | 2 - 3 | 165 | 60 | ||
AK004629.1 | - | 2 - 3 | 132 | 60 | ||
AK084385.1 | - | 2 - 3 | 125 | 60 | ||
AK088157.1 | - | 2 - 3 | 133 | 60 | ||
AK151239.1 | - | 2 - 3 | 132 | 60 | ||
AK173123.1 | - | 2 - 3 | 167 | 60 | ||
AK216037.1 | - | 2 - 3 | 74 | 60 | ||
BC083144.1 | - | 2 - 3 | 98 | 30432732 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
