Assay Details
| Gene Symbol: Ksr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013571.2 | NP_038599.1 | 15 - 16 | 2218 | 73 | |
| GenBank mRNA | AK137532.1 | - | 14 - 15 | 1955 | 73 | |
| U43585.1 | - | 15 - 16 | 2225 | 73 | ||
More Information
Set Membership: |
|

Ksr1
Genomic Map