Assay Details
| Gene Symbol: Sarm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168521.1 | NP_001161993.1 | 4 - 5 | 1687 | 68 | |
| NM_172795.3 | NP_766383.2 | 4 - 5 | 1687 | 68 | ||
| GenBank mRNA | AK172964.1 | - | 3 - 4 | 1950 | 68 | |
| AY444167.1 | - | 4 - 5 | 1391 | 68 | ||
| BC052098.1 | - | 4 - 5 | 1333 | 68 | ||
| BC058534.1 | - | 4 - 5 | 1634 | 6834056 | 68 | |
| BC080850.1 | - | 4 - 5 | 1687 | 6849138 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sarm1
Genomic Map