Assay Details
Gene Symbol: AW549877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145930.2 | NP_666042.2 | 2 - 3 | 249 | 156 | |
GenBank mRNA | AK045350.1 | - | 2 - 3 | 244 | 156 | |
AK154940.1 | - | 2 - 3 | 244 | 156 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
