Assay Details
| Gene Symbol: Cdh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007666.3 | NP_031692.2 | 1 - 2 | 123 | 116 | |
| GenBank mRNA | AK134611.1 | - | 1 - 2 | 160 | 116 | |
| CN704843.1 | - | 1 - 2 | 123 | 116 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK134611.1, NM_007666.3 transcripts.
Set Membership: |
|

Cdh6
Genomic Map