Assay Details
| Gene Symbol: Cdh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007666.3 | NP_031692.2 | 2 - 3 | 484 | 104 | |
| GenBank mRNA | AK134611.1 | - | 2 - 3 | 521 | 104 | |
| BC107008.1 | - | 1 - 2 | 225 | 40047565 | 104 | |
| BC107013.1 | - | 1 - 2 | 225 | 40048308 | 104 | |
| CN704843.1 | - | 2 - 3 | 484 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh6
Genomic Map