Assay Details
| Gene Symbol: Cdh6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007666.3 | NP_031692.2 | 7 - 8 | 1505 | 137 | |
| GenBank mRNA | BC107008.1 | - | 6 - 7 | 1246 | 40047565 | 137 |
| BC107013.1 | - | 6 - 7 | 1246 | 40048308 | 137 | |
| D82029.1 | - | 6 - 7 | 1246 | 137 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh6
Genomic Map