Assay Details
| Gene Symbol: Rps6kb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114334.1 | NP_001107806.1 | 11 - 12 | 1142 | 96 | |
| GenBank mRNA | AK012045.1 | - | 6 - 7 | 521 | 96 | |
| AK032724.1 | - | 11 - 12 | 1104 | 96 | ||
| AK135716.1 | - | 11 - 12 | 1110 | 96 | ||
| AK163258.1 | - | 11 - 12 | 1112 | 96 | ||
| AK214758.1 | - | 3 - 4 | 258 | 96 | ||
| BC038491.1 | - | 11 - 12 | 1115 | 4010710 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6kb1
Genomic Map