Assay Details
| Gene Symbol: Rps6kb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114334.1 | NP_001107806.1 | 14 - 15 | 1444 | 130 | |
| GenBank mRNA | AK012045.1 | - | 9 - 10 | 823 | 130 | |
| AK032724.1 | - | 14 - 15 | 1406 | 130 | ||
| AK135716.1 | - | 14 - 15 | 1412 | 130 | ||
| BC038491.1 | - | 14 - 15 | 1417 | 4010710 | 130 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rps6kb1
Genomic Map