Assay Details
| Gene Symbol: Zc3h7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081016.1 | NP_001074485.1 | 6 - 7 | 1030 | 85 | |
| GenBank mRNA | AK129268.1 | - | 6 - 7 | 707 | 85 | |
| AK147484.1 | - | 6 - 7 | 794 | 85 | ||
| AK147978.1 | - | 6 - 7 | 803 | 85 | ||
| AK165492.1 | - | 6 - 7 | 822 | 85 | ||
| AK171949.1 | - | 6 - 7 | 745 | 85 | ||
| BC059923.1 | - | 6 - 7 | 785 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zc3h7b
Genomic Map