Assay Details
| Gene Symbol: Mkl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082536.1 | NP_001076005.1 | 9 - 10 | 1468 | 114 | |
| NM_153049.3 | NP_694629.2 | 8 - 9 | 1230 | 114 | ||
| GenBank mRNA | AF385582.1 | - | 8 - 9 | 1001 | 114 | |
| AF532597.1 | - | 9 - 10 | 1352 | 114 | ||
| AK044188.1 | - | 9 - 10 | 1461 | 114 | ||
| AK089416.1 | - | 7 - 8 | 1060 | 114 | ||
| AK154561.1 | - | 9 - 10 | 1450 | 115 | ||
| AK155821.1 | - | 9 - 10 | 115 | |||
| AK155937.1 | - | 8 - 9 | 1214 | 114 | ||
| AK157985.1 | - | 7 - 8 | 1080 | 115 | ||
| BC050941.1 | - | 9 - 10 | 1361 | 6821370 | 114 | |
| BC054801.1 | - | 9 - 10 | 1404 | 114 | ||
| CA318154.1 | - | 3 - 4 | 495 | 114 | ||
| CX562326.1 | - | 1 - 2 | 130 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mkl1
Genomic Map