Assay Details
| Gene Symbol: Slc38a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175121.3 | NP_780330.2 | 10 - 11 | 1241 | 60 | |
| GenBank mRNA | AK031615.1 | - | 10 - 11 | 1272 | 60 | |
| AK129342.1 | - | 10 - 11 | 1202 | 60 | ||
| AK164568.1 | - | 4 - 5 | 5004 | 60 | ||
| BC037675.1 | - | 7 - 8 | 648 | 60 | ||
| BC041108.1 | - | 10 - 11 | 1111 | 4038546 | 60 | |
| BC048178.1 | - | 10 - 11 | 1241 | 6825801 | 60 | |
| BC049271.1 | - | 3 - 4 | 264 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a2
Genomic Map