Assay Details
| Gene Symbol: Slc38a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175121.3 | NP_780330.2 | 3 - 4 | 575 | 132 | |
| GenBank mRNA | AK031615.1 | - | 3 - 4 | 606 | 132 | |
| AK129342.1 | - | 3 - 4 | 536 | 132 | ||
| BC027259.1 | - | 3 - 4 | 479 | 132 | ||
| BC041108.1 | - | 3 - 4 | 445 | 4038546 | 132 | |
| BC048178.1 | - | 3 - 4 | 575 | 6825801 | 132 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a2
Genomic Map