Assay Details
| Gene Symbol: Kcnh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010601.3 | NP_034731.3 | 8 - 9 | 1502 | 88 | |
| GenBank mRNA | BC141013.1 | - | 8 - 9 | 1617 | 9055625 | 88 |
| BC145145.1 | - | 8 - 9 | 1617 | 9053696 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kcnh3
Genomic Map