Assay Details
| Gene Symbol: Ipo9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153774.1 | NP_722469.1 | 15 - 16 | 1921 | 82 | |
| GenBank mRNA | AF273672.1 | - | 15 - 16 | 1866 | 82 | |
| AF273673.1 | - | 15 - 16 | 1866 | 82 | ||
| AJ309238.1 | - | 15 - 16 | 1860 | 82 | ||
| AK036687.1 | - | 15 - 16 | 1871 | 82 | ||
| AK173127.1 | - | 15 - 16 | 1863 | 82 | ||
| AK211512.1 | - | 2 - 3 | 268 | 82 | ||
| BC098508.1 | - | 15 - 16 | 1871 | 30015513 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ipo9
Genomic Map