Assay Details
| Gene Symbol: Hadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008212.4 | NP_032238.2 | 1 - 2 | 230 | 90 | |
| GenBank mRNA | AK075609.1 | - | 1 - 2 | 157 | 90 | |
| AK132260.1 | - | 1 - 2 | 193 | 90 | ||
| AK148481.1 | - | 1 - 2 | 154 | 90 | ||
| AK148486.1 | - | 1 - 2 | 167 | 90 | ||
| AK167024.1 | - | 1 - 2 | 157 | 90 | ||
| AK168238.1 | - | 1 - 2 | 177 | 90 | ||
| AK168877.1 | - | 1 - 2 | 157 | 90 | ||
| AK169261.1 | - | 1 - 2 | 196 | 90 | ||
| BC028833.1 | - | 1 - 2 | 62 | 90 | ||
| BC064712.1 | - | 1 - 2 | 172 | 30303218 | 90 | |
| D29639.1 | - | 1 - 2 | 156 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hadh
Genomic Map