Assay Details
| Gene Symbol: Hadh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008212.4 | NP_032238.2 | 2 - 3 | 351 | 65 | |
| GenBank mRNA | AK075609.1 | - | 2 - 3 | 278 | 65 | |
| AK132260.1 | - | 2 - 3 | 314 | 65 | ||
| AK148481.1 | - | 2 - 3 | 275 | 65 | ||
| AK148486.1 | - | 2 - 3 | 288 | 65 | ||
| AK167024.1 | - | 2 - 3 | 278 | 65 | ||
| AK168238.1 | - | 2 - 3 | 298 | 65 | ||
| AK168877.1 | - | 2 - 3 | 278 | 65 | ||
| AK169261.1 | - | 2 - 3 | 317 | 65 | ||
| BC028833.1 | - | 2 - 3 | 183 | 65 | ||
| BC064712.1 | - | 2 - 3 | 293 | 30303218 | 65 | |
| D29639.1 | - | 2 - 3 | 277 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hadh
Genomic Map