Assay Details
| Gene Symbol: Wdfy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175546.4 | NP_780755.2 | 10 - 11 | 1295 | 87 | |
| GenBank mRNA | AK086275.1 | - | 10 - 11 | 1308 | 87 | |
| AK131958.1 | - | 10 - 11 | 1294 | 87 | ||
| AK151817.1 | - | 10 - 11 | 1251 | 87 | ||
| BC116632.1 | - | 10 - 11 | 1241 | 40056912 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Wdfy2
Genomic Map