Assay Details
| Gene Symbol: Ints6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008715.2 | NP_032741.2 | 7 - 8 | 1350 | 102 | |
| GenBank mRNA | BC059263.1 | - | 7 - 8 | 1350 | 6409580 | 102 |
More Information
Updated Mapping Information
Set Membership: |
|

Ints6
Genomic Map