Assay Details
| Gene Symbol: Ctsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007798.3 | NP_031824.1 | 2 - 3 | 252 | 107 | |
| GenBank mRNA | AK002456.1 | - | 2 - 3 | 201 | 107 | |
| AK083393.1 | - | 2 - 3 | 252 | 107 | ||
| AK147192.1 | - | 2 - 3 | 199 | 107 | ||
| AK149884.1 | - | 2 - 3 | 201 | 107 | ||
| AK151790.1 | - | 2 - 3 | 199 | 107 | ||
| AK159992.1 | - | 2 - 3 | 237 | 107 | ||
| AK167361.1 | - | 2 - 3 | 233 | 107 | ||
| AK170959.1 | - | 2 - 3 | 225 | 107 | ||
| AK190209.1 | - | 1 - 2 | 50 | 107 | ||
| AK191055.1 | - | 1 - 2 | 50 | 107 | ||
| AK191391.1 | - | 1 - 2 | 50 | 107 | ||
| AK196955.1 | - | 1 - 2 | 50 | 107 | ||
| AK198213.1 | - | 1 - 2 | 50 | 107 | ||
| AK201893.1 | - | 1 - 2 | 50 | 107 | ||
| AK202291.1 | - | 1 - 2 | 50 | 107 | ||
| AK203639.1 | - | 1 - 2 | 50 | 107 | ||
| BC006656.1 | - | 2 - 3 | 243 | 3500700 | 107 | |
| M14222.1 | - | 1 - 2 | 157 | 107 | ||
| S69034.1 | - | 2 - 3 | 248 | 107 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ctsb
Genomic Map