Assay Details
| Gene Symbol: Ctsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007798.3 | NP_031824.1 | 3 - 4 | 344 | 94 | |
| GenBank mRNA | AK002456.1 | - | 3 - 4 | 293 | 94 | |
| AK083393.1 | - | 3 - 4 | 344 | 94 | ||
| AK147192.1 | - | 3 - 4 | 291 | 94 | ||
| AK149884.1 | - | 3 - 4 | 293 | 94 | ||
| AK151790.1 | - | 3 - 4 | 291 | 94 | ||
| AK153047.1 | - | 3 - 4 | 290 | 94 | ||
| AK159992.1 | - | 3 - 4 | 329 | 94 | ||
| AK167361.1 | - | 3 - 4 | 325 | 94 | ||
| AK170959.1 | - | 3 - 4 | 317 | 94 | ||
| AK190209.1 | - | 2 - 3 | 142 | 94 | ||
| AK190539.1 | - | 2 - 3 | 142 | 94 | ||
| AK191055.1 | - | 2 - 3 | 142 | 94 | ||
| AK196955.1 | - | 2 - 3 | 142 | 94 | ||
| AK198213.1 | - | 2 - 3 | 142 | 94 | ||
| AK199268.1 | - | 2 - 3 | 143 | 94 | ||
| AK201893.1 | - | 2 - 3 | 142 | 94 | ||
| AK202291.1 | - | 2 - 3 | 94 | |||
| AK203639.1 | - | 2 - 3 | 142 | 94 | ||
| BC006656.1 | - | 3 - 4 | 335 | 3500700 | 94 | |
| M14222.1 | - | 2 - 3 | 249 | 94 | ||
| S69034.1 | - | 3 - 4 | 340 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ctsb
Genomic Map