Assay Details
| Gene Symbol: Slc25a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166450.2 | NP_001159922.1 | 1 - 2 | 153 | 94 | |
| NM_001290703.1 | NP_001277632.1 | 1 - 2 | 249 | 94 | ||
| NM_001290704.1 | NP_001277633.1 | 1 - 2 | 249 | 94 | ||
| NM_011398.3 | NP_035528.1 | 1 - 2 | 249 | 94 | ||
| GenBank mRNA | AF076981.1 | - | 1 - 2 | 249 | 94 | |
| AF535150.1 | - | 1 - 2 | 249 | 94 | ||
| AF535151.1 | - | 1 - 2 | 249 | 94 | ||
| CX244278.1 | - | 1 - 2 | 247 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF076981.1, AF535150.1, AF535151.1, NM_001166450.2, NM_001290703.1, NM_001290704.1, NM_011398.3 transcripts.
Set Membership: |
|

Slc25a14
Genomic Map