Assay Details
| Gene Symbol: Slc25a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166450.2 | NP_001159922.1 | 6 - 7 | 828 | 72 | |
| NM_001290703.1 | NP_001277632.1 | 8 - 9 | 1017 | 72 | ||
| NM_001290704.1 | NP_001277633.1 | 8 - 9 | 1008 | 72 | ||
| NM_001290705.1 | NP_001277634.1 | 5 - 6 | 583 | 72 | ||
| NM_011398.3 | NP_035528.1 | 6 - 7 | 915 | 72 | ||
| GenBank mRNA | AF076981.1 | - | 6 - 7 | 916 | 72 | |
| AF155812.1 | - | 5 - 6 | 503 | 72 | ||
| AF155813.1 | - | 5 - 6 | 494 | 72 | ||
| AF535150.1 | - | 8 - 9 | 1009 | 72 | ||
| AF535151.1 | - | 8 - 9 | 1018 | 72 | ||
| AK148277.1 | - | 6 - 7 | 2363 | 72 | ||
| BC048692.1 | - | 5 - 6 | 638 | 6333237 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a14
Genomic Map