Assay Details
| Gene Symbol: Slc25a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166450.2 | NP_001159922.1 | 8 - 9 | 1044 | 68 | |
| NM_001290703.1 | NP_001277632.1 | 10 - 11 | 1233 | 68 | ||
| NM_001290704.1 | NP_001277633.1 | 10 - 11 | 1224 | 68 | ||
| NM_001290705.1 | NP_001277634.1 | 7 - 8 | 799 | 68 | ||
| NM_011398.3 | NP_035528.1 | 8 - 9 | 1131 | 68 | ||
| GenBank mRNA | AF076981.1 | - | 8 - 9 | 1132 | 68 | |
| AF155812.1 | - | 7 - 8 | 719 | 68 | ||
| AF155813.1 | - | 7 - 8 | 710 | 68 | ||
| AF535150.1 | - | 10 - 11 | 1225 | 68 | ||
| AF535151.1 | - | 10 - 11 | 1234 | 68 | ||
| AK148277.1 | - | 8 - 9 | 2579 | 68 | ||
| BC048692.1 | - | 7 - 8 | 854 | 6333237 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a14
Genomic Map