Assay Details
| Gene Symbol: Rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009029.2 | NP_033055.2 | 20 - 21 | 2229 | 94 | |
| GenBank mRNA | AK132875.1 | - | 9 - 10 | 975 | 94 | |
| AK141028.1 | - | 20 - 21 | 2261 | 94 | ||
| AK146467.1 | - | 15 - 16 | 1560 | 94 | ||
| BC096525.1 | - | 20 - 21 | 2229 | 6832808 | 94 | |
| DQ400415.1 | - | 20 - 21 | 2079 | 94 | ||
| M26391.1 | - | 20 - 21 | 2177 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rb1
Genomic Map