Assay Details
| Gene Symbol: Rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009029.2 | NP_033055.2 | 22 - 23 | 2458 | 89 | |
| GenBank mRNA | AK132875.1 | - | 11 - 12 | 1204 | 89 | |
| AK141028.1 | - | 22 - 23 | 2490 | 89 | ||
| AK146467.1 | - | 17 - 18 | 1789 | 89 | ||
| BC096525.1 | - | 22 - 23 | 2458 | 6832808 | 89 | |
| DQ400415.1 | - | 22 - 23 | 2308 | 89 | ||
| M26391.1 | - | 22 - 23 | 2406 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rb1
Genomic Map