Assay Details
| Gene Symbol: Rb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009029.2 | NP_033055.2 | 4 - 5 | 632 | 131 | |
| GenBank mRNA | AK011246.1 | - | 4 - 5 | 632 | 131 | |
| AK141028.1 | - | 4 - 5 | 664 | 131 | ||
| BC096525.1 | - | 4 - 5 | 632 | 6832808 | 131 | |
| DQ400415.1 | - | 4 - 5 | 482 | 131 | ||
| M26391.1 | - | 4 - 5 | 580 | 131 | ||
More Information
Set Membership: |
|

Rb1
Genomic Map