Assay Details
Gene Symbol: Gpalpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026177.3 | NP_080453.2 | 5 - 6 | 666 | 95 | |
GenBank mRNA | AK004707.1 | - | 5 - 6 | 646 | 95 | |
AK082912.1 | - | 5 - 6 | 647 | 95 | ||
AK173238.1 | - | 5 - 6 | 611 | 95 | ||
BC029731.1 | - | 5 - 6 | 627 | 4946215 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
