Assay Details
| Gene Symbol: Nufip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013745.5 | NP_038773.1 | 2 - 3 | 514 | 107 | |
| GenBank mRNA | AF159549.1 | - | 2 - 3 | 472 | 107 | |
| AK009260.1 | - | 2 - 3 | 490 | 107 | ||
| AK032324.1 | - | 2 - 3 | 515 | 107 | ||
| AK156205.1 | - | 2 - 3 | 501 | 107 | ||
| BC020364.1 | - | 2 - 3 | 489 | 107 | ||
| BC056192.1 | - | 2 - 3 | 500 | 30024494 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nufip1
Genomic Map