Assay Details
| Gene Symbol: Cr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013499.2 | NP_038527.2 | 9 - 10 | 1547 | 93 | |
| GenBank mRNA | AI098173.1 | - | 1 - 2 | 93 | ||
| AK004825.1 | - | 9 - 10 | 1539 | 93 | ||
| BC028945.1 | - | 9 - 10 | 1314 | 5371665 | 93 | |
| BC092048.1 | - | 9 - 10 | 1305 | 3660242 | 93 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036250 | BC028945.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036250 | NM_013499.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036250 | AI098173.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2036250 | AK004825.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 769711 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 409572 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 632177 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 945930 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 58575 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 354444 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 965185 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75695 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 549435 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 460585 | BC020014.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 460585 | BC031731.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 719856 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 615759 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 94066 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 313723 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 384326 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 394215 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 261838 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 410490 | AK082248.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 45365 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561542 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 559649 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12448 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 405513 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99170 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 164478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 49041 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152911 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036250 | BC028945.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036250 | NM_013499.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036250 | AI098173.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2036250 | AK004825.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P295129 | AK017702.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P399580 | AK160440.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P399580 | BC028945.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P399580 | BC092048.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P399580 | M16179.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P399580 | M23446.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959588 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959596 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959594 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959630 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959591 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959592 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959590 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20959608 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04409728 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04409710 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04409774 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00290697 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152911 | - | - |
| Whole Mouse Genome G4122A | A_52_P399580 | AK160440.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P399580 | BC028945.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P399580 | BC092048.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P399580 | M16179.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P399580 | M23446.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P295129 | AK017702.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131332 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101982 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152911 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00366123 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00694561 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00694572 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00694560 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00694567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959608 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959623 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959619 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959630 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20959464 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959618 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959559 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959568 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959584 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00519537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959615 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00452809 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959605 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959590 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959574 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00362443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959621 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959558 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959613 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959625 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959561 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00694572 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959591 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959596 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959497 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00711093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959580 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00054650 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00241486 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00269466 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01020641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959594 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959610 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20959619 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10361285 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361287 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361286 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361290 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361289 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361291 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361283 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10361284 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1422563_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430131_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438920_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10361282 | - | - |
| Mouse Genome 430 2.0 Array | 1438920_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1430131_at | - | - |
| Mouse Genome 430 2.0 Array | 1438920_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1422563_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1422563_at | - | - |
| Mouse Genome 430 2.0 Array | 1430131_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12946_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12946_PM_at | - | - |
| Mouse Expression Set 430 | 1422563_PM_at | - | - |
| Mouse Expression Set 430 | 1422563_at | - | - |
| Mouse Expression Set 430 | 1430131_at | - | - |
| Mouse Expression Set 430 | 1438920_x_at | - | - |
| Mouse Expression Set 430 | 1430131_PM_at | - | - |
| Mouse Expression Set 430 | 1438920_PM_x_at | - | - |
| Murine Genome U74v2 Set | 101879_PM_s_at | - | - |
| Murine Genome U74v2 Set | 161648_PM_at | - | - |
| Murine Genome U74v2 Set | 161648_at | - | - |
| Murine Genome U74v2 Set | 101879_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422563_at | - | - |
| Mouse Genome 430A 2.0 Array | 1422563_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422563_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1422563_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438920_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438920_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430131_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1430131_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 380019 | - | - |
| MouseWG-6 v2 Expression BeadChip | 5550576 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 380019 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105910537 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5550576 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000380019 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005550576 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1230722 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2688629 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2424680 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1230722 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2688629 | - | - |
| mouse-6 v1.1 (Illumina) | scl0012946.2_160-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0012946.2_3-S | - | - |
| mouse-6 v1.1 (Illumina) | scl15731.3_14-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5550576 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 380019 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105910537 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0012946.2_3-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0012946.2_160-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105910537 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380019 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5550576 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2688631 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1234997 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234997 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2688631 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2424680 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0012946.2_3-S | - | - |

Cr1l
Genomic Map