Assay Details
| Gene Symbol: Fam228a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029107.2 | NP_083383.1 | 4 - 5 | 535 | 163 | |
| GenBank mRNA | AK015158.1 | - | 4 - 5 | 407 | 163 | |
| AK029492.1 | - | 4 - 5 | 536 | 163 | ||
| BC138846.1 | - | 4 - 5 | 470 | 8861868 | 163 | |
| BC138853.1 | - | 4 - 5 | 378 | 8861875 | 163 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam228a
Genomic Map