Assay Details
| Gene Symbol: Fam228a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029107.2 | NP_083383.1 | 5 - 6 | 616 | 81 | |
| GenBank mRNA | AK029492.1 | - | 5 - 6 | 617 | 81 | |
| AK133272.1 | - | 5 - 6 | 654 | 81 | ||
| BC138846.1 | - | 5 - 6 | 551 | 8861868 | 81 | |
| BC138853.1 | - | 5 - 6 | 459 | 8861875 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam228a
Genomic Map