Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400003938.mm.1 |
NM_027518.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003938.mm.1 |
NM_027518.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003938.mm.1 |
TC1400000479.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003939.mm.1 |
NM_027518.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003939.mm.1 |
NM_027518.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003939.mm.1 |
TC1400000479.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003933.mm.1 |
NM_027518.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003933.mm.1 |
NM_027518.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003933.mm.1 |
TC1400000479.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003934.mm.1 |
NM_027518.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003934.mm.1 |
NM_027518.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003934.mm.1 |
TC1400000479.mm.1 |
MTA TC |