Assay Details
| Gene Symbol: Psmc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025959.3 | NP_080235.2 | 10 - 11 | 802 | 110 | |
| GenBank mRNA | AK012174.1 | - | 10 - 11 | 803 | 110 | |
| AK144728.1 | - | 799 | 110 | |||
| AK166961.1 | - | 10 - 11 | 787 | 110 | ||
| BC025134.1 | - | 6 - 7 | 490 | 110 | ||
| BC037099.1 | - | 10 - 11 | 800 | 110 | ||
| BC043044.1 | - | 781 | 110 | |||
| BC057997.1 | - | 758 | 110 | |||
| BC116752.1 | - | 10 - 11 | 802 | 40126071 | 110 | |
| BC119168.1 | - | 10 - 11 | 802 | 8733917 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psmc6
Genomic Map