Assay Details
| Gene Symbol: Psmc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025959.3 | NP_080235.2 | 11 - 12 | 922 | 90 | |
| GenBank mRNA | AK008996.1 | - | 666 | 90 | ||
| AK012174.1 | - | 11 - 12 | 923 | 90 | ||
| AK014354.1 | - | 11 - 12 | 910 | 90 | ||
| AK144728.1 | - | 919 | 90 | |||
| AK166961.1 | - | 11 - 12 | 907 | 90 | ||
| BC025134.1 | - | 7 - 8 | 610 | 90 | ||
| BC037099.1 | - | 11 - 12 | 920 | 90 | ||
| BC043044.1 | - | 901 | 90 | |||
| BC057997.1 | - | 878 | 90 | |||
| BC116752.1 | - | 11 - 12 | 922 | 40126071 | 90 | |
| BC119168.1 | - | 11 - 12 | 922 | 8733917 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psmc6
Genomic Map