Assay Details
| Gene Symbol: Psmc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025959.3 | NP_080235.2 | 12 - 13 | 994 | 84 | |
| GenBank mRNA | AK008996.1 | - | 738 | 84 | ||
| AK012174.1 | - | 12 - 13 | 995 | 84 | ||
| AK014354.1 | - | 12 - 13 | 982 | 84 | ||
| AK144728.1 | - | 991 | 84 | |||
| AK166961.1 | - | 12 - 13 | 979 | 84 | ||
| BC025134.1 | - | 8 - 9 | 682 | 84 | ||
| BC037099.1 | - | 12 - 13 | 992 | 84 | ||
| BC043044.1 | - | 973 | 84 | |||
| BC057997.1 | - | 950 | 84 | |||
| BC116752.1 | - | 12 - 13 | 994 | 40126071 | 84 | |
| BC119168.1 | - | 12 - 13 | 994 | 8733917 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psmc6
Genomic Map