Assay Details
| Gene Symbol: Psmc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025959.3 | NP_080235.2 | 1 - 2 | 110 | 119 | |
| GenBank mRNA | AK012174.1 | - | 1 - 2 | 111 | 119 | |
| AK014354.1 | - | 1 - 2 | 98 | 119 | ||
| AK144728.1 | - | 107 | 119 | |||
| AK166961.1 | - | 1 - 2 | 95 | 119 | ||
| BC037099.1 | - | 1 - 2 | 108 | 119 | ||
| BC043044.1 | - | 89 | 119 | |||
| BC057997.1 | - | 66 | 119 | |||
| BC116752.1 | - | 1 - 2 | 110 | 40126071 | 119 | |
| BC119168.1 | - | 1 - 2 | 110 | 8733917 | 119 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psmc6
Genomic Map