Assay Details
| Gene Symbol: Psmc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025959.3 | NP_080235.2 | 2 - 3 | 193 | 162 | |
| GenBank mRNA | AK012174.1 | - | 2 - 3 | 194 | 162 | |
| AK014354.1 | - | 2 - 3 | 181 | 162 | ||
| AK144728.1 | - | 190 | 162 | |||
| AK166961.1 | - | 2 - 3 | 178 | 162 | ||
| BC037099.1 | - | 2 - 3 | 191 | 162 | ||
| BC043044.1 | - | 172 | 162 | |||
| BC057997.1 | - | 149 | 162 | |||
| BC116752.1 | - | 2 - 3 | 193 | 40126071 | 162 | |
| BC119168.1 | - | 2 - 3 | 193 | 8733917 | 162 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psmc6
Genomic Map