Assay Details
| Gene Symbol: Psmc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025959.3 | NP_080235.2 | 5 - 6 | 348 | 68 | |
| GenBank mRNA | AK008996.1 | - | 92 | 68 | ||
| AK012174.1 | - | 5 - 6 | 349 | 68 | ||
| AK014354.1 | - | 5 - 6 | 336 | 68 | ||
| AK144728.1 | - | 345 | 68 | |||
| AK166961.1 | - | 5 - 6 | 333 | 68 | ||
| BC025134.1 | - | 1 - 2 | 36 | 68 | ||
| BC037099.1 | - | 5 - 6 | 346 | 68 | ||
| BC043044.1 | - | 327 | 68 | |||
| BC057997.1 | - | 304 | 68 | |||
| BC116752.1 | - | 5 - 6 | 348 | 40126071 | 68 | |
| BC119168.1 | - | 5 - 6 | 348 | 8733917 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psmc6
Genomic Map