Assay Details
| Gene Symbol: Psmc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025959.3 | NP_080235.2 | 6 - 7 | 463 | 87 | |
| GenBank mRNA | AK008996.1 | - | 207 | 87 | ||
| AK012174.1 | - | 6 - 7 | 464 | 87 | ||
| AK144728.1 | - | 460 | 87 | |||
| AK166961.1 | - | 6 - 7 | 448 | 87 | ||
| BC025134.1 | - | 2 - 3 | 151 | 87 | ||
| BC037099.1 | - | 6 - 7 | 461 | 87 | ||
| BC043044.1 | - | 442 | 87 | |||
| BC057997.1 | - | 419 | 87 | |||
| BC116752.1 | - | 6 - 7 | 463 | 40126071 | 87 | |
| BC119168.1 | - | 6 - 7 | 463 | 8733917 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Psmc6
Genomic Map