Assay Details
| Gene Symbol: Psmc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025959.3 | NP_080235.2 | 8 - 9 | 611 | 99 | |
| GenBank mRNA | AK008996.1 | - | 355 | 99 | ||
| AK012174.1 | - | 8 - 9 | 612 | 99 | ||
| AK144728.1 | - | 608 | 99 | |||
| AK166961.1 | - | 8 - 9 | 596 | 99 | ||
| BC025134.1 | - | 4 - 5 | 299 | 99 | ||
| BC037099.1 | - | 8 - 9 | 609 | 99 | ||
| BC043044.1 | - | 590 | 99 | |||
| BC057997.1 | - | 567 | 99 | |||
| BC116752.1 | - | 8 - 9 | 611 | 40126071 | 99 | |
| BC119168.1 | - | 8 - 9 | 611 | 8733917 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Psmc6
Genomic Map